missing translation for 'onlineSavingsMsg'
Learn More

Robu Washing-Bottle, FC 13mm 500ml, Por. 4, GL 45

Brand:  Robu 42134

Washing-Bottle, FC 13mm 500ml, Por. 4, GL 45

Product Code. 10002121

  • 78.50 € / Each
Estimated Shipment: 16-10-2024
to see stock.
Add to basket
This item is not returnable. View return policy