Learn More
SCAT Waste Disposal Set, V2.0
The SCAT Waste Disposal Sets 1-4 are combinations of funnels and canisters for the safe disposal of liquid laboratory waste.
309.00 € - 595.00 €
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
17142576
|
SCAT
|
Set 1 |
309.00 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17132576
|
SCAT
307315 |
Set 2 |
450.00 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17162576
|
SCAT
307416 |
Set 3 |
595.00 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17152576
|
SCAT
307321 |
Set 4 |
337.00 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
Description
- Made of high quality and electrically conductive HDPE material
- With grounding cable to dissipate the electrical charge
- With splash guard and removable sieve for safe filling
- Sets consist of ball or lid funnels for individual applications
- 10 or 20 L disposal canisters with mechanical level indicators
- Can be optionally combined with collection tray
All canisters and funnels are connected via an S60/61 thread and their components are perfectly matched to each other.
Specifications
HPDE Electrostatic Conductive | |
S60/61 |