Learn More
Gosselin™ Waste and Sharps Container, White Hinged Cap
CORNING GOSSELIN™ Waste and Sharps Container, White Hinged Cap is constructed of polypropylene and available in a variety of sizes.
Brand:  Gosselin™ COX1100A-02
 View more versions of this product
Product Code. 15799709
  • 388.12 € / Pack of 60
Explore more special offers
Description
Description
  • Polypropylene construction
  • Suitable for waste and sharps
  • NFX 30-5111 compliant
  • Hinged cap
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.1 L
Hinged Top
Yellow
Polypropylene
NFX 30-5111
White
60/Cs.