Learn More
Rogo Sampaic™ Watch Glasses
Excellent for liquid evaporation, weighing solids, heating small amounts of substances and as beaker covers. Rogo Sampaic™ Watch Glasses are manufactured from durable clear borosilicate glass.
Specifications
Material Borosilicate Glass
Temperature Range (Metric) 500 °C
View More Specs
Products 9
Product Code Brand Diameter (Metric) Capacity (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Capacity (Metric) Price Quantity & Availability  
11702724
View Documents Promotion Details
Rogo Sampaic™
KBN013
60 mm 100 mL N/A
N/A
11792714
View Documents Promotion Details
Rogo Sampaic™
KBN011
40 mm 25 mL N/A
N/A
11738094
View Documents Promotion Details
Rogo Sampaic™
KBN017
120 mm 1000 mL N/A
N/A
11728094
View Documents Promotion Details
Rogo Sampaic™
KBN016
100 mm 400 to 600 mL N/A
N/A
11748094
View Documents Promotion Details
Rogo Sampaic™
KBN018
140 mm 2 L N/A
N/A
11724146
View Documents Promotion Details
Rogo Sampaic™
KBN019
160 mm 3 L N/A
N/A
11778084
View Documents Promotion Details
Rogo Sampaic™
KBN012
50 mm 50 mL N/A
N/A
11708094
View Documents Promotion Details
Rogo Sampaic™
KBN014
70 mm 150 mL N/A
N/A
11718094
View Documents Promotion Details
Rogo Sampaic™
KBN015
80 mm 250 mL N/A
N/A
Description
Description
  • Available in a range of diameters
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Borosilicate Glass
500 °C