missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, ACS Reagent, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
2.5 L
4 L
6 x 500 mL
4 x 2 L
4 x 4 L
Packaging:
Fiberboard Box
Glass Bottle
Poly Bottle
missing translation for 'unitSize'
2.5 L
4 L
500mL
Pack of 4
Pack of 6
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 100°C |
| UN Number | NONH for all modes of transport |
| Beilstein | 2050024 |
| Formula Weight | 18.02g/mol |
| Grade | ACS Reagent |
| Packaging | Poly Bottle |
| Density | 1.000g/mL at 3.98°C |
| Quantity | 500 mL |
| Chemical Name or Material | Water |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction