missing translation for 'onlineSavingsMsg'
Learn More

Water, ACS Reagent, Honeywell Riedel-de Haën™

Product Code. 15673630
Click to view available options
Quantity:
500 mL
2.5 L
4 L
6 x 500 mL
4 x 2 L
4 x 4 L
Packaging:
Fiberboard Box
Glass Bottle
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Melting Point 0°C
Boiling Point 100°C
UN Number NONH for all modes of transport
Beilstein 2050024
Formula Weight 18.02g/mol
Grade ACS Reagent
Packaging Poly Bottle
Density 1.000g/mL at 3.98°C
Quantity 6 x 500 mL
Chemical Name or Material Water
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'