missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Calibration and Matrix Blank For ICP, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep PLBLK-H2O-4L
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- DI Water Matrix Blank
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
ICP Water Blank | |
4 L | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Certified Reference Material | |
Plasma |
Triple-leached LDPE bottle | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Water Calibration and Matrix Blank For ICP, SPEX CertiPrep™ > 4L; Triple-leached LDPE bottle
Product intended for laboratory use only
Spot an opportunity for improvement?Share a Content Correction