missing translation for 'onlineSavingsMsg'
Learn More

Water, CHROMASOLV™, Honeywell Riedel-de Haën™

CHROMASOLV™, for pesticide residue analysis

71.87 € - 280.74 €

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15604190
View Documents Promotion Details
Honeywell-Chromasolv
34478-1L
1 L
71.87 €
1 L
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
 
15614190
View Documents Promotion Details
Honeywell-Chromasolv
34478-6X1L
6 x 1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
Specifications

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O

Specifications

7732-18-5
MFCD00011332
2050024
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02g/mol
1.000g/mL at 3.98°C
H2O
NONH for all modes of transport
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
O
18.015
CHEBI:15377
CHROMASOLV™
Water
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : ZC0110000

SDS
Documents

Documents

Product Certifications