missing translation for 'onlineSavingsMsg'
Learn More

Water, CHROMASOLV™, for HPLC, Honeywell Riedel-de Haën™

Product Code. 15612880
Click to view available options
Quantity:
1 L
2.5 L
4 L
6 x 1 L
Packaging:
Glass Bottle

CHROMASOLV™, for HPLC

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance max. 0.005 at 400 nm, max. 0.005 at 250 nm, max. 0.01 at 210 nm, max. 0.01 at 205 nm
Boiling Point 100°C
CAS Max % 100.0000%
Linear Formula H2O
Beilstein 2050024
Formula Weight 18.02
Grade HPLC
Packaging Glass Bottle
Density 1.000 g/mL at 3.98 °C(lit.)
Quantity 6 x 1 L
Chemical Name or Material Water, CHROMASOLV™, for HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.