missing translation for 'onlineSavingsMsg'
Learn More

Water, Demineralized, Chem-Lab

Product Code. p-8014440
Click to view available options
Quantity:
10 L
25 L
Unit Size:
10 L
25 L
This item is not returnable. View return policy

Product Code. 11969242

Brand: Chem-Lab CL02.2102.9010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for laboratory use. Water, Demineralized, Chem-Lab is purified to remove minerals and salts with LF <5μS. There are less than 5ppm of non-volatile matter. Available in 10 and 25L cartons.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH ±7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.016g/mol
Odor Odorless
Filtered Through LF <5μS
Density 1.00g/mL
Quantity 10 L
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.