missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, DNA Grade, DNASE, Protease free, Fisher BioReagents
CAS: 7732-18-5 | H2O | 18.015 g/mol
Brand: Fisher BioReagents BP2470-1
36.39 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
This item is not returnable.
View return policy
Description
Nalgene Poly Bottle; 1L
DNA-grade Water is DNase- and Protease-free and is suitable for all DNA work.
Specifications
Water | |
7 | |
Colorless | |
100°C | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
DNA | |
17.5mmHg at 20°C | |
1.000g/cm³ | |
Liquid |
For DNA Work, DNASE, Protease free | |
7732-18-5 | |
0°C | |
DNase free | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Nalgene Poly Bottle | |
Filtered through a 0.2-micron filter and autoclaved. | |
1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Water, DNA Grade, DNASE, Protease free, Fisher BioReagents > Nalgene Poly Bottle; 1L
Spot an opportunity for improvement?Share a Content Correction