missing translation for 'onlineSavingsMsg'
Learn More

Water, (DNASE, RNASE free), Fisher BioReagents™

Product Code. p-23724
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Nalgene Poly Bottle
Poly Bottle
Unit Size:
100mL
50mL
This item is not returnable. View return policy

Product Code. 10295243

Brand: Fisher BioReagents BP248450

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 100mL, Vapor Pressure: 17.5mmHg at 20 deg.C, 100 deg.C, CHEBI:15377, Gray, Colorless, 1.000g/cm3, DNase free, Filtered through a 0.2-micron filter., 18.02, Low hazard for usual industrial or commercial handling.

Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
DNase DNase free
Merck Index 15, 10237
Formula Weight 18.02
Packaging Nalgene Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.2-micron filter.
Treatment(s) DEPC-treated and autoclaved.
Density 1.000g/cm³
Quantity 50 mL
Physical Form Liquid
Chemical Name or Material Water, DNASE, RNASE free
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.