missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Water, Endotoxin-free Product Code.: 15466979

Thermo Scientific Chemicals Water, Endotoxin-free

Product Code. 15466979
1 L
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy

Product Code. 15466979

Brand: Thermo Scientific Chemicals J65589.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Water Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Water Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.

Solubility
Miscible with methanol, acetone and ethanol.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxideShow MoreShow Less
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Melting Point 32°F
Boiling Point 100°C
Beilstein 2050024
Merck Index 1410039
Solubility Information Miscible with methanol,acetone and ethanol.
Formula Weight 18.02
Grade Endotoxin-Free
Odor Odorless
Density 1
Quantity 1 L
Refractive Index 1.34
Physical Form Liquid
Chemical Name or Material Water, Endotoxin-free
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14L562 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog NumberJ65589.AP,J65589.K2
Lot NumberN14L563 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog NumberJ65589.AP,J65589.K2
Lot NumberY18K565 Certificate TypeCertificate of Analysis Date02/12/2024 Catalog NumberJ65589.AP,J65589.K2
Lot NumberW26K561 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog NumberJ65589.AP,J65589.K2
Lot NumberW28K522 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog NumberJ65589.AP,J65589.K2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Water, Endotoxin-free >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.