missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, Extra Pure, Deionized, Thermo Scientific Chemicals
61.41 € - 165.00 €
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10347071
|
Thermo Scientific Acros
345470050 |
5 L | Plastic Drum |
61.41 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11339986
|
Thermo Scientific Acros
345470250 |
25 L | Plastic Drum |
165.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Specifications
7732-18-5 | |
100.0°C | |
H2O | |
16,383 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Extra Pure | |
0.89 mPa.s (25°C) | |
1.3334 | |
Liquid |
Colorless | |
5.0μS/cm Max. | |
H2O | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
25ppm max. | |
1.0000g/mL | |
1 | |
Water |
Safety and Handling
Product Identifier
- Water
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Water, Extra Pure, Deionized, Thermo Scientific Chemicals