missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Water For Injection (WFI) for Cell Culture
GREENER_CHOICE

High quality water for use as a solvent in the preparation of cell culture media and laboratory reagents.

20.16 € - 194.00 €

Specifications

pH 4 to 6
Grade Cell Culture
Sterility Sterile-filtered
Treatment(s) None
Purification Method Distilled and filtered (0.1 μm)
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10378939
GREENER_CHOICE
View Documents
Gibco™
A1287301
500 mL
20.16 €
Each
Please to purchase this item. Need a web account? Register with us today!
10282830
GREENER_CHOICE
View Documents
Gibco™
A1287302
1000 mL
36.43 €
Each
Please to purchase this item. Need a web account? Register with us today!
12093835
GREENER_CHOICE
View Documents
Gibco™
A1287303
10 x 500 mL
187.00 €
5000mL
Please to purchase this item. Need a web account? Register with us today!
10690861
GREENER_CHOICE
View Documents
Gibco™
A1287304
6 x 1000 mL
194.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere
  • The water is prepared by distillation and filtration (0.1 μm), with no added substances
  • In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances
  • Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly
  • It can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
  • Should be stored at 15° to 30°C
Specifications

Specifications

4 to 6
Sterile-filtered
Distilled and filtered (0.1 μm)
Gibco™
Cell Culture
None
Room Temperature
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture

SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Water For Injection (WFI) for Cell Culture

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.