missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Water For Injection (WFI) for Cell Culture

Gibco™ Water For Injection (WFI) for Cell Culture is high-quality water for use as a solvent in the preparation of cell culture media, laboratory reagents, buffers, and bioprocess liquids.

362.00 € - 1590.00 €

Specifications

pH 4 to 6
Grade USP/EP
Packaging Universal Bag
Sterility Sterile-filtered
Treatment(s) None
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17840925
View Documents
Gibco™
A1287305
20 L
362.00 €
20 L
Please to purchase this item. Need a web account? Register with us today!
17880925
View Documents
Gibco™
A1287306
200 L
1590.00 €
200 L
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

4 to 6
Universal Bag
None
Room Temperature
USP/EP
Sterile-filtered
Membrane-Filtered
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 months from date of manufacture

Recommended Storage : Store at 2°C to 30°C

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Water For Injection (WFI) for Cell Culture

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.