missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, for HPLC gradient grade, Thermo Scientific Chemicals
34.20 € - 58.70 €
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10596261
|
Thermo Scientific Acros
268300010 |
1 L |
34.20 €
1 L |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
10043370
|
Thermo Scientific Acros
268300025 |
2.5 L |
58.70 €
2.50 L |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCChemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
7732-18-5 | |
100.0°C | |
H2O | |
16,383 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
HPLC Gradient | |
0.0003% max. | |
0.89 mPa.s (25°C) | |
1.3334 | |
Water, For HPLC gradient grade |
Colorless | |
H2O | |
MFCD00011332 | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Glass Bottle | |
17.5mmHg at 20°C | |
1.0000g/mL | |
1 |
Safety and Handling
missing translation for 'einecsNumber' : 231-791-2
missing translation for 'rtecsNumber' : ZC0110000
missing translation for 'tsca' : TSCA
RUO – Research Use Only