missing translation for 'onlineSavingsMsg'
Learn More

Water, Microbial Cell Culture Grade, Fisher BioReagents

65.15 €

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11373892
View Documents
Fisher BioReagents
BP2820-1
1 L Nalgene Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Ultrapure water for preparing microbiological growth media.
Specifications

Chemical Identifiers

7732-18-5
18.015
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane

Specifications

7732-18-5
100°C
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Microbial Cell Culture
0.03 Micron filtered and autoclaved
Liquid
Colorless
2μS/cm Max.
0.025 EU/mL max.
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.01
17.5mmHg at 20°C
1.000g/cm³
Water, 0.03μm filtered and autoclaved
Videos
Safety and Handling

Safety and Handling

Low hazard for usual industrial or commercial handling.

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Rinse with water. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . .

NFPA

Health:0
Flammability:0
Instability:0

Product Identifier
  • Water

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.