missing translation for 'onlineSavingsMsg'
Learn More

Water, Optima™ LC/MS Grade, Fisher Chemical™

Product Code. 10728098
1 L
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.

Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number 11332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance ≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm
pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Merck Index 15, 10237
Formula Weight 18.02
Grade LC-MS & UHPLC-UV
Packaging Wide Neck Amber Bottle
Residue after Evaporation ≤1ppm
Vapor Pressure 17.5mmHg at 20°C
Filtered Through 0.03 micron filtered.
Density 1.000g/cm³
Certificate of Analysis Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, Optima™ LC/MS Grade, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.