Learn More
3M™ Clean-Trace™ Water Positive Control HWATP10
Designed to provide assurance that your 3M hygiene monitoring reagents and instruments are being stored and used correctly.
Brand:  3M™ Clean-Trace™ 7000086277
Encompass
Product Code. 17273734
  • POA / Pack of 10
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Verifies the performance of reagents and instruments
  • Can be used as a quench test with 3M™ Clean-Trace™ Water ATP tests
Specifications
Specifications
Microbiology Media
30 ng