missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Aqua-Soft System PG 8597

Aqua-Soft system PG 8597 - Twin-tank water softener for the continuous removal of soft water up to 40 °dH

Brand:  Miele Professional™ 6943120

Product Code. 15400585

This item is not returnable. View return policy

Description

Description

  • Output: continuous 19 l/min, max. 30 l/min
  • Freestanding appliance with rollers, top loaded
  • 1 container for 20 kg salt, 2 containers for 4.5 l resin
  • Volume controlled two-chamber system, current-less operation
Specifications

Specifications

White
Water Pre-treatment
360 mm
360 mm
Washer-disinfectors
Plastic
570 mm
19 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers