Learn More
Elgas™ PURELAB flex 1 Water Dispenser
Elgas™ PURELAB flex 1 Water Dispenser is designed to simply and elegantly dispense water when it is connected to a reservoir or distribution loop.
Brand:  Elgas™ PRIPLB0160
Product Code. 13115970
  • 11050.00 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to deliver accuracy, flexibility, and ease-of-use.
  • Works as a dispenser as well as a simple deionization system.
  • Customized settings to suit your applicaion provide flexibility and control.
  • Easy access facilitates routine maintenance.
  • Fully recirculating.
  • Integrated filtration.
  • Data capture system allows you to download system and performance validation via USB.
  • Ideally suited for dispensing, with DI pack, as well as general laboratory applications requiring type 2 water.
Specifications
Specifications
Daily volume: <100 L
900 to 1020 mm
374 mm
236 mm
374 x 236 x 900-1020 mm
374mm
10 kg
Up to 2.0 L/min.