missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Water Purification Systems Storage Reservoirs

Safely store purified water from your Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ system with one of these compatible storage reservoirs.

Brand:  Thermo Scientific™ 06.5084

 View more versions of this product

Product Code. 11307101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Safely store purified water from a compatible system with Thermo Scientific™ Water Purification Systems Storage Reservoirs. For use with Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ systems, these reservoirs are compatible with the following models: GenPure, GenPure Pro, GenPure xCAD, GenPure xCAD Plus, Pacific RO, Pacific TII and Smart2Pure.

Features:
  • Large opening offers easy and effective cleaning.
  • Optional safety filters include a CO2 absorber prevents conductivity increases from drawn-in CO2 and a sterile overflow to prevent microorganisms from entering and contaminating reservoir water.
  • Polyethylene reservoir is constructed with inert materials opaque to light.
  • Recirculation pump (optional) protects water from bacterial growth during standstill while maintaining a low conductivity value.
  • Wall mount is optional.
  • Conical bottom outlet allows for complete draining.
For use with GenPure, GenPure Pro, and GenPure xCAD purification systems only:
  • 06.5038
  • 06.5068
For use with Pacific RO water purification systems only:
  • 06.5034
  • 06.5064
  • 06.5084
For use with Pacific TII water purification systems only:
  • 06.5032
  • 06.5062
  • 06.5082
For use with Pacific RO & Pacific TII water purification systems only:
  • 06.5033
  • 06.5063
  • 06.5083
For use with Smart2Pure 12 water purification systems only:
  • 06.5040
  • 06.5070
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 L
Storage Reservoir, with level display and pressure pump
Pacific RO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.