missing translation for 'onlineSavingsMsg'
Learn More

Water, for qPCR or RT-qPCR, Fisher BioReagents

Product Code. 15822544
Click to view available options
Quantity:
1 L
10 L
Packaging:
Poly Bottle
Poly CUBE

High purity water rigorously tested for degrading enzymes and genomic DNA that interefere with amplification and can lead to false-positive signals in PCR, qPCR and RT-qPCR applications.

  • Minimize the risk of false positive signals
  • Tested for DNase, RNase, protease, nickase, and E. coli and human genomic DNA using sensitive assays
  • Evaluated for metal impurities at low ppb concentrations to ensure optimal enzymatic activity, especially for polymerase during PCR
  • Sub-micron filtered to remove particulates
  • Suitable for use in qPCR and RT-qPCR for genomics work, microarrays, and other sensitive life science applications
  • Also useful for the manufacturing of PCR-related kits and mastermixes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 5.4 to 7.0
Color Clear, Colorless
Melting Point 0°C
Boiling Point 100°C
Conductivity ≤2μS/cm
DNase Not detected
Endotoxin <0.1EU/mL
Formula Weight 18.02
Grade qPCR
Packaging Poly CUBE
Quantity 10 L
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.