missing translation for 'onlineSavingsMsg'
Learn More

Water, Reagent (Deionized water), ACS, Thermo Scientific Chemicals

42.70 € - 152.76 €

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11378078
View Documents
Thermo Scientific Alfa Aesar
036645.K2
1 L
46.13 €
1 L
In Stock
to see stock.
Add to basket
 
11368078
View Documents
Thermo Scientific Alfa Aesar
036645.K2
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15421467
View Documents
Thermo Scientific Alfa Aesar
036645.M1
4 x 1 L
152.76 €
1 L
In Stock
to see stock.
Add to basket
 
11388078
View Documents
Thermo Scientific Alfa Aesar
036645.K7
4 L
91.86 €
4 L
In Stock
to see stock.
Add to basket
 
Description

Description

Used in conductometric titration, Synthesis of nanomaterials. Mostly it is used as solvent in industries and in all research fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O

Specifications

7732-18-5
0°C
H2O
dihydrogen oxide, dihydrogen monoxide
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Odorless
1.333
Water
Colorless
100°C
MFCD00011332
Miscible in methanol,acetone,Ethanol.
O
18.015
CHEBI:15377
ACS
1
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-791-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.