missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
31.50 € - 57.00 €
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10674432
|
Thermo Scientific Acros
326650010 |
1 L | Glass Bottle |
31.50 €
1 L |
In Stock
Log in to see stock. |
||||
10031561
|
Thermo Scientific Acros
326650025 |
2.5 L | Glass Bottle |
57.00 €
2.50 L |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
7732-18-5 | |
H2O | |
MFCD00011332 | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
0.0002% max. | |
1.0000g/mL | |
1 |
100.0°C | |
H2O | |
16,383 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Residue Analysis | |
0.89 mPa.s (25°C) | |
1.3334 | |
Water |
Safety and Handling
missing translation for 'einecsNumber' : 231-791-2
missing translation for 'rtecsNumber' : ZC0110000
missing translation for 'tsca' : TSCA
RUO – Research Use Only