missing translation for 'onlineSavingsMsg'
Learn More
Water, RNase, DNase-free Product Code.: 15403359

Water, RNase, DNase-free

Product Code. 15403359
3.8 L
Click to view available options
Quantity:
3.8 L
This item is not returnable. View return policy

Product Code. 15403359

Brand: Thermo Scientific Alfa Aesar J60610.=C

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7732-18-5 | H2O | 18.015 g/mol

Water, RNase, DNase-free high quality, RNase/DNase-free water is used in all molecular biology applications like PCR, DNA. Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Water, RNase, DNase-free high quality, RNase/DNase-free water is used in all molecular biology applications like PCR, DNA. Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and i
TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5
Color Colorless
Boiling Point 100°C
Molecular Formula H2O
MDL Number MFCD00011332
Beilstein 2050024
Merck Index 14,10039
Synonym dihydrogen oxide, dihydrogen monoxide
Solubility Information It is soluble in water.
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.02
Odor Odorless
Density 1.000 g/mL
Quantity 3.8 L
Physical Form Liquid
Chemical Name or Material Water, RNase, DNase-free
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, RNase, DNase-free >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.