missing translation for 'onlineSavingsMsg'
Learn More

Water, RNase-Free, DEPC Treated, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

34.20 € - 87.10 €

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15875398
View Documents
Thermo Scientific
J70783.AC
25 mL Nalgene Bottle
34.20 €
25mL
In Stock
to see stock.
Edge
Add to basket
 
15885398
View Documents
Thermo Scientific
J70783.AE
100 mL Nalgene Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15895398
View Documents
Thermo Scientific
J70783.AP
500 mL Nalgene Bottle
67.20 €
500mL
In Stock
to see stock.
Add to basket
 
15805408
View Documents
Thermo Scientific
J70783.K2
1 L Nalgene Bottle
87.10 €
1 L
In Stock
to see stock.
Add to basket
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O

Specifications

7732-18-5
100°C
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
1
Liquid
0°C
H2O
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
O
18.015
CHEBI:15377
Molecular Biology
1.000
Water
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-791-2

missing translation for 'rtecsNumber' : ZC0110000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.