Learn More
Brady™ Water Soluble Paper Label Tape for M6/M7 Printers
This B-403 material of these labels completely dissolves in warm water within 30 seconds, leaving no adhesive residue behind.
Brand:  Brady™ 173536
 View more versions of this product
Product Code. 17869114
  • 119.00 € / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for use in laboratory settings, asset tracking or general use
  • Easy to remove, environmentally responsible, print-on-demand THT label solution
  • Easy removal saves time during re-labeling process
  • Pressure-sensitive adhesive allows for quick and easy application
  • Made of B-403 paper material
Specifications
Specifications
Not Autoclavable
M6C-750-403
None
Matte
Paper
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
50 ft.
-94°F to +212°F
0.003 in.
50 ft. x 0.75 in.
15.24 m
19.05 mm
ASTM D 1000, ASTM D 2979, RoHS Compliant to 2005/618/EC MCV Amendment to RoHS Directive 2002/95/EC
Acrylic
White
Continuous
Single-side Printable
Labels
Asset Tracking, General Identification, Laboratory Labeling
No
Rectangle
-70°C to +100°C
0.0762 mm
50 ft.
0.75 in.