Learn More
Brady™ Water Soluble Paper Labels with Ribbon
For the ultimate solution in label and adhesive removal, choose Brady’s B-403 Water Dissolvable Label. Ideal for laboratories, temporary identification needs, work-in-process product flows, and other general identification applications.
Brand: Brady™ 152248
This item is not returnable.
View return policy
Description
- For the ultimate solution in label and adhesive removal, choose Brady’s B-403 Water Dissolvable Label.
- Ideal for laboratories, temporary identification needs, work-in-process product flows, and other general identification applications.
- Work-In-Process: warm water and 30 seconds is all it takes to completely remove the label and adhesive.
- Application Versatility: From laboratories to restaurants, this dissolvable paper material is an excellent solution for temporary identification of any kind.
- Black print on white paper with easy to apply pressure-sensitive adhesive.
- Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).
- For material specific performance and properties, please refer to the Technical Data Sheet (TDS).
Specifications
Label | |
Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-PLUS Label Printer, BMP21 LAB Label Printer | |
Thermal Transfer | |
Acrylic | |
Matte |
Black on White | |
Paper (B-403) | |
25 ft. | |
Non-sterile |