missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, for UHPLC Gradient Grade Analysis, Fisher Chemical™
23.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
- Efficiency: lot-to-lot consistency ensures reproducible results
- Low acidity/alkalinity levels avoid interactions with samples
- Peak shape: low baseline noise at 210nm and 254nm improves peak shape
- Speed: filtered to 0.1μm, allowing analysis to be performed faster at higher pressures with less risk of pump blockages
- Sensitivity: interference-free with outstandingly high UV transmission for improved resolution and sensitivity
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless |
Boiling Point | 100°C |
Formula Weight | 18.02g/mol |
Packaging | Plastic Coated Glass Bottle |
Vapor Pressure | 17.5mmHg at 20°C |
Quantity | 1 L |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'