missing translation for 'onlineSavingsMsg'
Learn More

Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™

Product Code. 11934391
Click to view available options
Quantity:
500 mL
1 L
2 L
Packaging:
LDPE Bottle

Packaged in specially precleaned LDPE bottles to protect product integrity

  • Evaluated for 65 metal impurities in ppt range
  • Also specified for ≤1ppb for chloride, phosphate and sulfate anion impurities
  • Packaged in precleaned LDPE bottles to maintain product integrity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number 11332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade Ultra Trace Elemental Analysis
Packaging LDPE Bottle
Vapor Pressure 17.5mmHg at 20°C
Density 1.000g/cm³
Certificate of Analysis Certificate of Lot Analysis included with each bottle.
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'