missing translation for 'onlineSavingsMsg'
Learn More

Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™

Product Code. p-216448 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2 L
Packaging:
LDPE Bottle
Unit Size:
1 L
2 L
500mL
This item is not returnable. View return policy

Product Code. 11934391

Brand: Fisher Chemical undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Packaged in specially precleaned LDPE bottles to protect product integrity

  • Evaluated for 65 metal impurities in ppt range
  • Also specified for ≤1ppb for chloride, phosphate and sulfate anion impurities
  • Packaged in precleaned LDPE bottles to maintain product integrity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number 11332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade Ultra Trace Elemental Analysis
Packaging LDPE Bottle
Vapor Pressure 17.5mmHg at 20°C
Density 1.000g/cm³
Certificate of Analysis Certificate of Lot Analysis included with each bottle.
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.