Learn More
Lab Armor™ Waterless Bead Bath without Beads
Low maintenance and eco-friendly alternative to water baths. These Waterless Bead Baths use metallic beads instead of water, delivering exceptional temperature uniformity.
Brand: Lab Armor™ 10122-17
2218.39 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
- Low maintenance
- Eco-friendly - beads are recyclable
- Use 50% less electricity
- No germicides needed
- Beads hold things in place, eliminating the need for racks and weights
- Bath is always ready, eliminating warm-up times
- Safer operation than oil baths
- Dry, inert, non-toxic aluminum oxide ensures hazard-free operation emptying, cleaning, and refilling
- Beads have high thermal conductivity: ±180°C
- Beads measure 5 to 8 mm dia.
Specifications
Waterless bead bath | |
14 L | |
Heating a variety of samples and containers, including multi-well plates, Petri dishes and open-top samples at any angle. Beads can also be used in a container for freezers, ovens and incubators. | |
220 V |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.