missing translation for 'onlineSavingsMsg'
Learn More

Lab Armor™ Waterless Bead Bath

Low maintenance and eco-friendly alternative to water baths. These Waterless Bead Baths use metallic beads instead of water, delivering exceptional temperature uniformity.

2630.00 € - 3320.00 €

Specifications

Description Waterless bead bath
For Use With (Application) Heating a variety of samples and containers, including multi-well plates, Petri dishes and open-top samples at any angle. Beads can also be used in a container for freezers, ovens and incubators.
Voltage 220 V
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15311296
View Documents
Lab Armor™
10122-05
5 L
2630.00 €
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
 
15630034
View Documents
Lab Armor™
10122-07
6 L
3320.00 €
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
 
Description

Description

  • Low maintenance
  • Eco-friendly - beads are recyclable
  • Use 50% less electricity
  • No germicides needed
  • Beads hold things in place, eliminating the need for racks and weights
  • Bath is always ready, eliminating warm-up times
  • Safer operation than oil baths
  • Dry, inert, non-toxic aluminum oxide ensures hazard-free operation emptying, cleaning, and refilling
  • Beads have high thermal conductivity: ±180°C
  • Beads measure 5 to 8 mm dia.
Note: Beads are not included; sold separately.

Specifications

Specifications

Waterless bead bath
Heating a variety of samples and containers, including multi-well plates, Petri dishes and open-top samples at any angle. Beads can also be used in a container for freezers, ovens and incubators.
220 V
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.