Learn More
Labconco™ WaterPro™ PS Water Polishing Station
Filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
Brand:  Labconco™ 9047101
 View more versions of this product
Product Code. 15492212
  • 760.00 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
General Chemistry/UF Polishing Kit includes Activated Carbon Cartridge (1), polishing grade mixed bed Deionization Cartridges (3). Kit is required for models 9000520, 9000521, 9000530, 9000531, 9000540, 9000541, 9000560, 9000561, 9000570, 9000571, 9000580, 9000581, 9000720, 9000721, 9000730, 9000731, 9000740, 9000741, 9000760, 9000761, 9000770, 9000771, 9000780 and 9000781).
†Comes with valve to dispense Type I water and gun to dispense Type I, pyrogen-free water.
Order Info
All models require Start-Up Kits, sold separately. Unit dimensions (including bowls): 7 3/4L x 31 1/8W x 28 7/8 in.H (20 x 79 x 73cm). For 115V 60Hz; 230V 50Hz models also available through your Fisher Scientific™ Representative.
Specifications
Specifications
WaterPro PS and PS/UF Startup Kit
WaterPro™ PS polishing station
WaterPro PS and PS/UF
4.3 kg
PS and PS/UF Startup Kit
Activated Carbon cartridge (1) and polishing grade mixed bed deionization cartridges (3)
9.5 lb.