missing translation for 'onlineSavingsMsg'
Learn More
Kolmi Hopen™ Waterproof CPE Shoecovers Product Code.: 17730159
Add to basket

Kolmi Hopen™ Waterproof CPE Shoecovers

Product Code. 17730159
38 cm
Click to view available options
Thickness:
38 cm
41 cm
This item is not returnable. View return policy

Product Code. 17730159

Brand: Kolmi Hopen™ H10100

Add to basket
Add to basket
This item is not returnable. View return policy

Wear these CPE shoecovers in the industrial or catering settings.

  • Disposable and waterproof
  • Intended to be worn by visitors, patients or caregivers
  • Can be used as an object of environmental control in HACCP approaches and various industries
  • H10150 model is thicker and more resistant; can be used in static work situations

Specifications

Size Medium
Material CPE
Color Blue
Disposable Single-use
Thickness 38 cm
For Use With (Application) Industrial
Certifications/Compliance ISO 9001
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kolmi Hopen™ Waterproof CPE Shoecovers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.