Learn More
Hanna Instruments™ Waterproof pH/Conductivity/TDS Tester
Makes measuring pH quick and easy. Hanna Instruments™ Waterproof pH/Conductivity/TDS Tester is lightweight and waterproof offering high accuracy pH, EC/TDS and temperature measurements in a single handheld unit.
Brand:  Hanna Instruments™ HI98129
Product Code. 10381032
  • 372.00 € / Each
Edge
Explore more special offers
Description
Description
  • Combo testers designed for high accuracy measurements using one meter
  • Easy to read dual LCD, pH/°C or TDS/°C
  • Automatic temperature compensation
  • Automatic calibration
  • 'Hold' function
  • Automatic shut off
  • Replaceable pH electrode
Specifications
Specifications
±2%
±2%
Yes
LCD
0.00 to 14.00
0.01
HI98129
1ppm (mg/L)
±0.5°C
32.0°F to 140.0°F
±0.05
100 hr.
Automatic, at one or two points with two sets of standard buffers (pH 4.01 / 7.01 / 10.01 or pH 4.01
0 to 3999 μS/cm
0 to 2000 ppm (mg/L)
0.0°C to 60.0°C
1μS/cm
0 to 2000ppm (mg/L)
pH: automatic; EC/TDS: automatic with β adjustable from 0.0 to 2.4% /°C
0.1 / 0.1°C/°F
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products