missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wax Ceresin, Technical, White, Fisher Chemical
Click to view available options
250g, Plastic powder jar
Specifications
Specifications
CAS | 8001-75-0 |
Melting Point | 61°C |
Color | White to Yellow |
Packaging | Plastic powder jar |
Molecular Formula | C30H50O2 |
MDL Number | 146361 |
InChI Key | DSEKYWAQQVUQTP-UHFFFAOYNA-N |
Molecular Weight (g/mol) | 442.73 |
Physical Form | Solid |
Safety and Handling
Product Identifier
- WAX CERESIN WHITE
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction