Learn More
Thermo Scientific™ WebSeal™ Sealing Tapes
Product Code. p-6153756
Click to view available options
Material:
Aluminum, Silicone
PES, Silicone
PET, Silicone
Polyethylene, Silicone
Polyolefin, Rubber
Color:
Clear
Silver
Unit Size:
Pack of 100
Product Code. 15122719
Brand: Thermo Scientific™ 60180M142
Only null left
Add to basket
Add to basket
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes.
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes. Available in a selection of thin, lightweight materials and adhesives to meet the needs of any application or assay, they minimize evaporation and protect samples from contamination and spilling.
Easy, Consistent Sealing
  • Pressure-sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Tapes seal onto the plate with a convenient, handheld applicator (available separately)
TRUSTED_SUSTAINABILITY
Especificaciones
Color Clear
Material Polyethylene, Silicone
No. per Pack 100
Product Type Non-sterile Sealing Tape
Product Line Webseal
Adhesive Pressure Sensitive
Temperature Range (Metric) -40°C to +90°C
Plate Compatibility All Microplates
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado