Learn More
Microspec™ Wedge Shaped Spreaders
Wedge shaped to reduce tendency to flip
Brand:  Microspec™ D10
 View more versions of this product
Product Code. 15712125
  • 27.65 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Reduces tendency to flip
Specifications
Specifications
Single-use
7.1 in.
Microseal Pouch
High-impact Polystyrene
Aseptically Manufactured
Blue
180 mm
25 Pouches of 20
Spreader