Learn More
Medicom WeePro Labo Overboots
Click to view available options
One Size
Description
- New material MICROMIUM™, light and very low contaminant
- Cleanroom apparel compatible from ISO class 4
- Sealed packaging by pair with an Easy Tear system
- Overpack by 25 pairs
- Apparel sterilized by gamma irradiation
Specifications
Specifications
Size | Universal |
Material | Microporous Fabric |
Latex Free | No |
Color | White |
Disposable | Single-use |
Sterility | Sterile |
Closure Type | Elastic Top with Ankle Ties |
Sole Type | Slip-resistant |
Product Type | Overboots |