Learn More
Nabertherm™ Weighing Furnace, 1100 LT Series
Designed especially for combustion loss determination in the laboratory. The included software records both the temperature and the weight loss.
Brand:  Nabertherm™ LW095H1ANO
 View more versions of this product
Product Code. 17348019
  • 10730.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Maximum Temperature: 1100°C
  • Heating from two sides
  • Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Dual shell housing made of sheets of textured stainless steel
  • Optional lift door with hot surface facing away from the operator
  • Adjustable working air inlet in the door
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low noise operation
  • Delivery includes base, ceramic plunger with base plate in the furnace lining, precision scale and software package
  • 4 scales available for different maximum weights and scaling ranges
  • Process control and documentation for temperature and combustion loss via VCD software package for monitoring, documentation and control
  • Defined application within the constraints of the operating instructions
Specifications
Specifications
C550
1100°C
455 x 415 x 980 mm
9 L
240 x 230 x 170 mm
35 kg