missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Weighing Papers

Nitrogen-free for a very smooth surface and good sample recovery.

Brand:  Fisherbrand™

44.67 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 15630951

  • 59.05 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

 

  • Strong enough to support dense material
  • 500 per pack
  • Disposable
  • 76.2 x 76.2mm

 

Specifications

Specifications

76.2 x 76.2 mm
500/Pk.
Weighing Paper
100 mm
100 mm
Square
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.