missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 908 Weighing Papers

Smooth paper with satin appearance and wetstrength resistance.

Brand:  Fisherbrand™ 0908F00003

 View more versions of this product

Product Code. 11744576

  • 30.65 € / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Smooth paper 45g/m2
  • With satin appearance, wetstrength resistance
Specifications

Specifications

Weighing of all types of substances such as beet mash, powders and liquids
3.7 in.
Paper
110 mm
Rectangular
95 x 110mm (3.7 x 4.3 in.)
95 mm
4.3 in.
Weighing Paper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.