Learn More
Sartorius™ Weighing Tables
Prevent damage to the surface of weights with these weighing tables.
Brand:  Sartorius™ YWT09
2380.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11353284
  • 2665.00 € / Each
Explore more special offers
Description
Description
  • High density of the table's solid material provides excellent support for balances to meet the highest weighing requirements such as those stipulated for calibration of weights using mass comparators
  • Recommended if cleanliness of the weighing area is a major requirement
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Wood with cast stone inset
750 x 900 x 750 mm
Weighing Table