Learn More
Welch™ DirecTorr™ Gold Synthetic Pump Oil
Designed for excellent resistance when pumping corrosive gases or vapors
221.00 € - 296.00 €
Specifications
Product Type | Gold Vacuum Pump Oil |
---|---|
For Use With (Application) | Oil enables long service life and superior vacuum pump performance in applications where corrosive gases and vapors may be ingested |
For Use With (Equipment) | Recommended for ChemStar pump, belt-driven or direct-drive vacuum pumps |
Quantity | 1 Ea. |
Product Code | Brand | Volume (Metric) | Volume (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Volume (English) | Price | Quantity & Availability | ||||
12671095
|
Welch™
U8995G-15 |
3.78 L | 1 gal. | N/A | |||||
12819558
|
Welch™
U8995G-15 |
3.78 L | 1 gal. |
296.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
Description
Designed for excellent resistance when pumping corrosive gases or vapors
Enables excellent vacuum performance in both belt- and direct-driven vacuum pumps under harsh condition. Synthetic base stock has no aromatic compounds or sulfur. This purity inhibits varnishing, sludging and carbon buildup when pumping corrosives.
- Double-distilled synthetic hydrocarbon oil
- High stability in chemical environments with exceptionally low vapor pressure
- Miscible with conventional hydrocarbon oils and can be used without rebuilding the pump
- No additives or inhibitors
Specifications
Gold Vacuum Pump Oil | |
Oil enables long service life and superior vacuum pump performance in applications where corrosive gases and vapors may be ingested | |
Recommended for ChemStar pump, belt-driven or direct-drive vacuum pumps | |
1 Ea. |