Learn More
Thermo Scientific™ Wellwash™ Microplate Washer
Wash 96-well plates securely with this easy-to-use, reliable microplate-strip washer for routine ELISA applications.
6210.00 € - 6280.00 €
Specifications
Accuracy | < 5% @ 300μL |
---|---|
Certifications/Compliance | CE Marked |
Connections | USB |
Wash/Waste Bottle Capacity | 1 |
Bottles | Wash Bottle (2 L), Waste Bottle (2 L) |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
11708016
|
Thermo Scientific™
5165000 |
Wellwash 1x8 |
6210.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15174725
|
Thermo Scientific™
5165040 |
Wellwash 1x12 |
6280.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Wash 96-well plates securely with the Thermo Scientific™ Wellwash™ Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications. The Wellwash features a graphical user interface, local language versions and USB port for optimal ease-of-use and convenience.
Simplify your ELISA workflow by using our eGuide to see our complete portfolio for all your ELISA needs.
Features:
- Large color clear display and keypad for easy, convenient use
- User interface is logic and intuitive for quick protocols development
- Easy-to-use software requires minimal training for fast startup
- Unpressurized bottles are safe and secure to use
- Liquid level sensors are present in both wash and waste bottle for security
- Automatic prime feature assure safe performance
- Aerosol cover prevents aerosolization of infectous diseases
- Plate sensor recognizes if plate is present
- Wellwash is designed to efficiently remove the excess liquid from the wells
- Low residual volumes provide optimal washing performance and reliable assay results
Includes:
1x2L wash bottle, 1x2L waste bottle, 1x8L wash head, aerosol cover
Recommended for:
ELISA-based assays
Warranty: 1 year
Specifications
< 5% @ 300μL | |
USB | |
Wash Bottle (2 L), Waste Bottle (2 L) | |
50 to 400 μL | |
<2 μL | |
50 to 1000 μL | |
ELISA-Based Assays | |
No | |
385 mm | |
345 cm | |
240 mm | |
17.6 lb. | |
100/240 V |
CE Marked | |
1 | |
< 3% (CV) @ 300μL (96-well plate) | |
96-Well Plates | |
8 kg | |
LCD | |
50/60 Hz | |
15.2 in. | |
13.6 in. | |
9.4 in. | |
No | |
100/240 V 50/60 Hz | |
5 to 100 mL |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.