Learn More
IBA Lifesciences WET FRED Set
The WET FRED Set is an application aid for large volumes of lysates or culture supernatants
Brand:  IBA Lifesciences 2-0911-001
 View more versions of this product
Product Code. 15583067
  • 138.00 € / 1 set
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • WET FRED enables the application of large cell culture supernatant or lysate volumes to a gravity flow column in a simple way
  • Based on the siphon principle, it connects the gravity flow column with the bottle containing culture supernatant, lysate, or buffer that are to be applied
Specifications
Specifications
1 mL Gravity Flow Columns
1 Set
Eases Protein Purification
Protein Purification Aid