missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ 0.8 μm Pore Size (ME 27) Mixed Cellulose Ester Membranes, Plain

Whatman™ (ME 27) Mixed Cellulose Ester Membranes, Plain are particularly effective in applications requiring higher flow rates and larger volume filtration including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.

123.13 € - 407.00 €

Specifications

Autoclavable Autoclavable
Bubble Point 1.3 bar
Filter Type Plain
Material MCE
Material (Membrane) ME 27
View More Specs

Products 3
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
11395014
View Documents
Cytiva
10400921
100 mm
407.00 €
Pack of 50
In Stock
to see stock.
Add to basket
 
11305024
View Documents
Cytiva
10400906
25 mm
123.13 €
Pack of 100
In Stock
to see stock.
Add to basket
 
11325024
View Documents
Cytiva
10400914
50 mm
165.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
Description

Description

  • Material: Mixed Cellulose Ester
  • Material (Membrane): ME 27
  • Filter Type: Plain
  • Pore Size: 0.8 μm
  • Autoclavable: Yes
  • Sterility: Non-sterile
  • Bubble Point: 1.3 bar
  • Flow Rate: 2.8 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
  • Gas Flow Rate: 11.6 sec./100 mL (at Δp = 3 mbar)
  • For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications

Specifications

Autoclavable
Plain
ME 27
Non-sterile
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Plain
1.3 bar
MCE
0.8 μm
2.8 sec./100 mL/12.5 cm⊃ (at Δp = 0.9 bar)
11.6 sec./100 mL (at Δp = 3 mbar)
Videos
Safety and Handling

Safety and Handling

Signal Word
  • ||Warning

Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Product Certifications